Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOXL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002318.2 | NP_002309.1 | 2 | 566 | ||
GenBank mRNA | AK222477.1 | - | 2 | 477 | ||
AK312266.1 | - | 2 | 478 | |||
BC000594.2 | - | 2 | 436 | 3347512 | ||
BC020481.1 | - | 2 | 486 | |||
KF928961.1 | - | 1 | 227 | |||
KF975332.1 | - | 1 | 239 | |||
U89942.1 | - | 2 | 474 |