Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VCL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003373.3 | NP_003364.1 | 2, 3 | 331 | ||
NM_014000.2 | NP_054706.1 | 2, 3 | 331 | |||
GenBank mRNA | AK126978.1 | - | 2, 3 | 586 | ||
AK296509.1 | - | 2, 3 | 327 | |||
AK300280.1 | - | 2, 3 | 329 | |||
AL708715.1 | - | 2, 3 | 331 | |||
BC039174.1 | - | 2, 3 | 318 | 4520338 | ||
BX537994.1 | - | 4, 5 | 196 | |||
EU794673.1 | - | 2, 3 | 331 | |||
M33308.1 | - | 2, 3 | 287 |