Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAM18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190956.1 | NP_001177885.1 | 2 | 154 | ||
NM_001320313.1 | NP_001307242.1 | 2 | 154 | |||
NM_014237.2 | NP_055052.1 | 2 | 154 | |||
NR_135201.1 | 2 | 154 | ||||
GenBank mRNA | AJ133004.1 | - | 2 | 99 | ||
AK225744.1 | - | 2 | 91 | |||
AK313961.1 | - | 2 | 144 | |||
AY358321.1 | - | 2 | 141 | |||
BC034624.1 | - | 2 | 134 | |||
BC070279.1 | - | 2 | 137 | |||
BC121045.1 | - | 2 | 99 | 40117918 | ||
BC121046.1 | - | 2 | 99 | |||
DB046002.1 | - | 2 | 154 | |||
DB049415.1 | - | 2 | 154 |