Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMACR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167595.1 | NP_001161067.1 | 4, 5 | 843 | ||
NM_014324.5 | NP_055139.4 | 4, 5 | 843 | |||
NM_203382.2 | NP_976316.1 | 3, 4 | 682 | |||
GenBank mRNA | AF047020.1 | - | 4, 5 | 835 | ||
AF158378.1 | - | 4, 5 | 812 | |||
AJ130733.1 | - | 4, 5 | 835 | |||
AK000912.1 | - | 4, 5 | 828 | |||
AK222831.1 | - | 4, 5 | 815 | |||
AK225749.1 | - | 4, 5 | 852 | |||
AK292988.1 | - | 3, 4 | 632 | |||
AL545355.3 | - | 4, 5 | 754 | |||
AY935981.1 | - | 4, 5 | 756 | |||
BC009471.1 | - | 3, 4 | 598 | 2958112 | ||
BQ941482.1 | - | 2, 3 | 352 | |||
EF560721.1 | - | 4, 5 | 766 |
Gene Symbol: C1QTNF3-AMACR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037951.1 | 8, 9 | 1103 | |||
GenBank mRNA | AB209261.1 | - | 8, 9 | 1112 |