Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACAD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014384.2 | NP_055199.1 | 3 | 422 | ||
XM_005271505.3 | XP_005271562.1 | 3 | 430 | |||
XM_011542750.2 | XP_011541052.1 | 3 | 430 | |||
XM_017017542.1 | XP_016873031.1 | 3 | 430 | |||
XM_017017543.1 | XP_016873032.1 | 3 | 430 | |||
XM_017017544.1 | XP_016873033.1 | 3 | 430 | |||
XM_017017545.1 | XP_016873034.1 | 3 | 430 | |||
XM_017017548.1 | XP_016873037.1 | 3 | 430 | |||
XM_017017549.1 | XP_016873038.1 | 3 | 430 | |||
GenBank mRNA | AB209538.1 | - | 2 | 217 | ||
AF126245.1 | - | 3 | 449 | |||
AK000359.1 | - | 3 | 394 | |||
AK299492.1 | - | 2 | 313 | |||
AK299795.1 | - | 3 | 404 | |||
AK301518.1 | - | 2 | 190 | |||
AK309838.1 | - | 3 | 425 | |||
BC001964.1 | - | 3 | 391 | 3462406 | ||
BI549961.1 | - | 3 | 427 |