Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SENP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267594.1 | NP_001254523.1 | 4, 5 | 542 | ||
NM_001267595.1 | NP_001254524.1 | 4, 5 | 797 | |||
XM_006719361.3 | XP_006719424.1 | 4, 5 | 677 | |||
XM_011538244.2 | XP_011536546.1 | 4, 5 | 717 | |||
XM_011538245.2 | XP_011536547.1 | 3, 4 | 629 | |||
XM_017019229.1 | XP_016874718.1 | 5, 6 | 815 | |||
XM_017019230.1 | XP_016874719.1 | 5, 6 | 770 | |||
XM_017019231.1 | XP_016874720.1 | 5, 6 | 715 | |||
XM_017019232.1 | XP_016874721.1 | 5, 6 | 442 | |||
XM_017019233.1 | XP_016874722.1 | 4, 5 | 725 | |||
XM_017019234.1 | XP_016874723.1 | 4, 5 | 635 | |||
XM_017019235.1 | XP_016874724.1 | 4, 5 | 368 | |||
XM_017019236.1 | XP_016874725.1 | 4, 5 | 642 | |||
XM_017019237.1 | XP_016874726.1 | 4, 5 | 542 | |||
GenBank mRNA | AF149770.1 | - | 2, 3 | 217 | ||
AK292060.1 | - | 4, 5 | 313 | |||
AK312874.1 | - | 4, 5 | 542 | |||
AL582855.3 | - | 4, 5 | 664 | |||
BC045639.1 | - | 4, 5 | 613 | 5298667 |