Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAMR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001991.2 | NP_001001991.1 | 5 | 963 | ||
NM_001282675.1 | NP_001269604.1 | 7 | 953 | |||
NM_015430.3 | NP_056245.2 | 5 | 963 | |||
GenBank mRNA | AF370388.1 | - | 4 | 451 | ||
AK027841.1 | - | 5 | 563 | |||
AK297092.1 | - | 7 | 954 | |||
AL832391.1 | - | 5 | 529 | |||
BC089434.1 | - | 5 | 555 | 30558617 | ||
BX640676.1 | - | 5 | 963 |