Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKD2L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253837.1 | NP_001240766.1 | 2 | 689 | ||
NM_016112.2 | NP_057196.2 | 2 | 667 | |||
GenBank mRNA | AF053316.1 | - | 2 | 159 | ||
AF073481.1 | - | 2 | 667 | |||
BC025665.1 | - | 2 | 364 | 5200440 | ||
DQ084244.1 | - | 2 | 143 |