Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKD2L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253837.1 | NP_001240766.1 | 4 | 928 | ||
NM_016112.2 | NP_057196.2 | 4 | 906 | |||
XM_011540323.2 | XP_011538625.1 | 5 | 469 | |||
XM_011540325.2 | XP_011538627.1 | 6 | 579 | |||
XM_017016875.1 | XP_016872364.1 | 5 | 417 | |||
XM_017016876.1 | XP_016872365.1 | 4 | 400 | |||
GenBank mRNA | AF053316.1 | - | 4 | 398 | ||
AF073481.1 | - | 4 | 906 | |||
AF092170.1 | - | 2 | 185 | |||
AF094827.1 | - | 3 | 238 | |||
BC025665.1 | - | 4 | 603 | 5200440 | ||
DQ084244.1 | - | 4 | 382 |