Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LTF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199149.1 | NP_001186078.1 | 2 | 295 | ||
NM_001321121.1 | NP_001308050.1 | 2 | 406 | |||
NM_001321122.1 | NP_001308051.1 | 5 | 639 | |||
NM_002343.5 | NP_002334.2 | 2 | 406 | |||
GenBank mRNA | AF332168.1 | - | 2 | 113 | ||
AK093852.1 | - | 5 | 642 | |||
AK290859.1 | - | 2 | 151 | |||
AK292813.1 | - | 2 | 151 | |||
AK298035.1 | - | 2 | 295 | |||
AK303889.1 | - | 2 | 151 | |||
AK316330.1 | - | 2 | 215 | |||
AY137470.1 | - | 2 | 113 | |||
AY165046.1 | - | 2 | 125 | |||
AY178998.2 | - | 2 | 113 | |||
AY360320.1 | - | 2 | 114 | |||
AY493417.1 | - | 2 | 223 | |||
AY875691.1 | - | 2 | 113 | |||
BC015822.1 | - | 2 | 155 | 4251222 | ||
BC015823.1 | - | 2 | 157 | 4294752 | ||
BC022347.1 | - | 2 | 151 | 4251058 | ||
BP342557.1 | - | 2 | 354 | |||
CD722125.1 | - | 2 | 139 | |||
DQ522304.1 | - | 2 | 113 | |||
EU779935.1 | - | 2 | 113 | |||
EU794669.1 | - | 2 | 148 | |||
KF992208.1 | - | 2 | 125 | |||
KT006756.1 | - | 2 | 113 | |||
M83202.1 | - | 2 | 120 | |||
M83205.1 | - | 2 | 99 | |||
M93150.1 | - | 2 | 130 | |||
U07643.1 | - | 2 | 210 | |||
X52941.1 | - | 2 | 107 | |||
X53961.1 | - | 2 | 407 |