Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LUM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002345.3 | NP_002336.1 | 2 | 649 | ||
GenBank mRNA | AK223177.1 | - | 2 | 334 | ||
AK312682.1 | - | 2 | 352 | |||
BC007038.1 | - | 2 | 401 | 3950745 | ||
BC035997.1 | - | 2 | 323 | 4688606 | ||
BT006707.1 | - | 1 | 260 | |||
U18728.1 | - | 2 | 340 | |||
U21128.1 | - | 2 | 344 |