Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LY6E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127213.1 | NP_001120685.1 | 3 | 294 | ||
NM_002346.2 | NP_002337.1 | 3 | 299 | |||
GenBank mRNA | AF085875.1 | - | 1 | 35 | ||
AK129615.1 | - | 3 | 256 | |||
AK223074.1 | - | 3 | 278 | |||
AK311983.1 | - | 3 | 255 | |||
BC003392.1 | - | 3 | 203 | |||
BC119708.1 | - | 2 | 180 | 40117169 | ||
BC119709.1 | - | 2 | 180 | 40117170 | ||
CB138313.1 | - | 3 | 288 | |||
DB458093.1 | - | 3 | 299 | |||
U42376.1 | - | 3 | 234 | |||
U56145.1 | - | 2 | 140 | |||
Z68179.1 | - | 3 | 234 |