Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017414.3 | NP_059110.2 | 11 | 1858 | ||
XM_006724074.2 | XP_006724137.1 | 11 | 1929 | |||
GenBank mRNA | AF176642.2 | - | 10 | 1540 | ||
AJ243526.1 | - | 11 | 1722 | |||
AL136690.1 | - | 11 | 1679 | |||
BC014896.1 | - | 11 | 1664 | 3910720 |