Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLK15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277081.1 | NP_001264010.1 | 5 | 977 | ||
NM_001277082.1 | NP_001264011.1 | 4 | 840 | |||
NM_017509.3 | NP_059979.2 | 5 | 980 | |||
NR_102274.1 | 5 | 859 | ||||
XM_006723265.3 | XP_006723328.1 | 6 | 1781 | |||
XM_011527085.2 | XP_011525387.1 | 6 | 1777 | |||
XM_011527087.2 | XP_011525389.1 | 5 | 1530 | |||
XM_011527089.2 | XP_011525391.1 | 5 | 1651 | |||
GenBank mRNA | AF303046.1 | - | 5 | 979 | ||
AY373373.1 | - | 4 | 1069 | |||
AY373374.1 | - | 4 | 932 |