Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OGDHL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018245.2 | NP_060715.2 | 3 | 464 | ||
XM_011539946.1 | XP_011538248.1 | 3 | 349 | |||
XM_011539947.1 | XP_011538249.1 | 3 | 555 | |||
XM_017016401.1 | XP_016871890.1 | 3 | 428 | |||
XM_017016402.1 | XP_016871891.1 | 3 | 339 | |||
XM_017016403.1 | XP_016871892.1 | 2 | 203 | |||
GenBank mRNA | AB033116.3 | - | 2 | 327 | ||
AK001713.1 | - | 3 | 408 | |||
AK225558.1 | - | 3 | 395 | |||
AK290226.1 | - | 2 | 437 | |||
AK307759.1 | - | 2 | 223 | |||
BC026320.1 | - | 3 | 383 | 4797814 | ||
DA238469.1 | - | 3 | 464 |