Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018397.4 | NP_060867.2 | 8 | 1796 | ||
XM_006713250.3 | XP_006713313.1 | 8 | 1578 | |||
XM_006713251.3 | XP_006713314.1 | 8 | 1755 | |||
XM_006713252.3 | XP_006713315.1 | 7 | 1435 | |||
XM_011533938.2 | XP_011532240.1 | 8 | 1612 | |||
XM_011533939.2 | XP_011532241.1 | 8 | 2451 | |||
XM_017006797.1 | XP_016862286.1 | 8 | 1609 | |||
XM_017006798.1 | XP_016862287.1 | 8 | 1601 | |||
XM_017006799.1 | XP_016862288.1 | 7 | 1432 | |||
GenBank mRNA | AJ272267.1 | - | 6 | 1020 | ||
AK055402.1 | - | 1 | 442 | |||
AK297575.1 | - | 4 | 479 | |||
BC034502.1 | - | 8 | 1554 | 4824572 |