Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNJ16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270422.1 | NP_001257351.1 | 6 | 1217 | ||
NM_001291622.1 | NP_001278551.1 | 6 | 1171 | |||
NM_001291623.1 | NP_001278552.1 | 3 | 964 | |||
NM_001291624.1 | NP_001278553.1 | 4 | 1092 | |||
NM_001291625.1 | NP_001278554.1 | 5 | 1173 | |||
NM_018658.2 | NP_061128.2 | 5 | 1089 | |||
NM_170741.2 | NP_733937.2 | 4 | 1008 | |||
NM_170742.2 | NP_733938.2 | 4 | 1045 | |||
XM_005257337.4 | XP_005257394.1 | 3 | 891 | |||
XM_006721885.3 | XP_006721948.1 | 7 | 1297 | |||
XM_006721886.3 | XP_006721949.1 | 6 | 1169 | |||
XM_006721887.3 | XP_006721950.1 | 6 | 1162 | |||
XM_011524781.2 | XP_011523083.1 | 5 | 1128 | |||
XM_017024609.1 | XP_016880098.1 | 7 | 1400 | |||
XM_017024610.1 | XP_016880099.1 | 6 | 1135 | |||
XM_017024611.1 | XP_016880100.1 | 6 | 1209 | |||
XM_017024612.1 | XP_016880101.1 | 5 | 1128 | |||
XM_017024613.1 | XP_016880102.1 | 6 | 1256 | |||
GenBank mRNA | AF153815.1 | - | 5 | 1029 | ||
AF153816.1 | - | 5 | 1029 | |||
AF153817.1 | - | 3 | 801 | |||
AF179353.1 | - | 1 | 543 | |||
AK225944.1 | - | 8 | 1054 | |||
AK290799.1 | - | 4 | 898 | |||
BC033038.2 | - | 4 | 1035 | 5262150 |