Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPCAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002354.2 | NP_002345.2 | 1, 2 | 438 | ||
GenBank mRNA | AK026585.1 | - | 1, 2 | 275 | ||
BC014785.1 | - | 1, 2 | 262 | 3861826 | ||
BP262680.1 | - | 1, 2 | 434 | |||
CR542259.1 | - | 1, 2 | 80 | |||
CR542283.1 | - | 1, 2 | 80 | |||
DA963872.1 | - | 1, 2 | 275 | |||
DB044092.1 | - | 1, 2 | 444 | |||
M26481.1 | - | 1, 2 | 239 | |||
M32306.1 | - | 1, 2 | 101 | |||
M32325.1 | - | 1, 2 | 234 | |||
M33011.1 | - | 1, 2 | 187 | |||
X14758.1 | - | 1, 2 | 234 |