Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNPEP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319182.1 | NP_001306111.1 | 4 | 576 | ||
NM_001319183.1 | NP_001306112.1 | 3 | 716 | |||
NM_001319184.1 | NP_001306113.1 | 3 | 716 | |||
NM_020216.3 | NP_064601.3 | 4 | 865 | |||
XM_005245421.1 | XP_005245478.1 | 3 | 383 | |||
XM_017002022.1 | XP_016857511.1 | 3 | 404 | |||
GenBank mRNA | AJ242586.1 | - | 4 | 866 | ||
AJ296161.1 | - | 4 | 836 | |||
AK026644.1 | - | 1 | 68 | |||
AK097372.1 | - | 5 | 779 | |||
AL390139.1 | - | 3 | 399 | |||
BC001064.2 | - | 4 | 850 | 2823082 | ||
BC012166.1 | - | 4 | 842 | 4636934 | ||
BY994108.1 | - | 4 | 553 | |||
DA359487.1 | - | 3 | 354 |