Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DECR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020664.3 | NP_065715.1 | 9 | 1095 | ||
GenBank mRNA | AJ293009.1 | - | 9 | 1010 | ||
AK055839.1 | - | 10 | 1741 | |||
AK094340.1 | - | 4 | 1579 | |||
AK128012.1 | - | 10 | 1247 | |||
AK225961.1 | - | 9 | 1264 | |||
AL442076.1 | - | 2 | 222 | |||
BC010740.1 | - | 9 | 1081 | 3877040 | ||
BC011968.1 | - | 9 | 1000 | 3866503 |