Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020903.2 | NP_065954.1 | 4 | 2935 | ||
GenBank mRNA | AF229438.1 | - | 1 | 2612 | ||
AK308285.1 | - | 4 | 3033 | |||
BC130394.1 | - | 2 | 2695 | 40146425 | ||
BC130398.1 | - | 2 | 2695 | 40146429 |