Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020935.2 | NP_065986.2 | 4 | 531 | ||
XM_005246720.2 | XP_005246777.1 | 4 | 603 | |||
XM_005246721.3 | XP_005246778.1 | 4 | 495 | |||
XM_005246722.3 | XP_005246779.1 | 4 | 535 | |||
XM_005246724.1 | XP_005246781.1 | 4 | 531 | |||
XM_011511538.2 | XP_011509840.1 | 3 | 476 | |||
XM_011511541.2 | XP_011509843.1 | 4 | 531 | |||
XM_017004577.1 | XP_016860066.1 | 4 | 489 | |||
GenBank mRNA | AK095577.1 | - | 4 | 489 | ||
AK128583.1 | - | 4 | 535 | |||
AL832645.2 | - | 4 | 530 | |||
BC050461.1 | - | 4 | 500 | |||
BC133007.1 | - | 4 | 416 | 40147029 | ||
BC133009.1 | - | 4 | 461 | 40147031 | ||
BC144249.1 | - | 4 | 416 | 9052769 | ||
BC144252.1 | - | 4 | 456 | 9052772 | ||
BX538024.1 | - | 4 | 496 |