Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCPEP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021626.2 | NP_067639.1 | 7, 8 | 714 | ||
XM_005257557.2 | XP_005257614.1 | 7, 8 | 696 | |||
GenBank mRNA | AF113214.1 | - | 7, 8 | 639 | ||
AF282618.1 | - | 7, 8 | 693 | |||
AK027373.1 | - | 7, 8 | 687 | |||
AK298161.1 | - | 7, 8 | 633 | |||
AY358559.1 | - | 7, 8 | 694 | |||
BC072405.1 | - | 7, 8 | 664 | 6137668 |