Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134223.1 | NP_001127695.1 | 3 | 564 | ||
NM_001286767.1 | NP_001273696.1 | 3 | 452 | |||
NM_001286768.1 | NP_001273697.1 | 4 | 596 | |||
NM_022832.3 | NP_073743.2 | 3 | 488 | |||
XM_017008557.1 | XP_016864046.1 | 3 | 454 | |||
GenBank mRNA | AK022614.1 | - | 3 | 308 | ||
AK024318.1 | - | 3 | 429 | |||
AK296493.1 | - | 4 | 569 | |||
AK299883.1 | - | 3 | 365 | |||
AK302438.1 | - | 3 | 563 | |||
BC037574.1 | - | 3 | 437 | 5529667 | ||
CN483539.1 | - | 3 | 488 | |||
GU455414.1 | - | 3 | 303 |