Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134223.1 | NP_001127695.1 | 9 | 2566 | ||
NM_001286767.1 | NP_001273696.1 | 9 | 2454 | |||
NM_001286768.1 | NP_001273697.1 | 10 | 2657 | |||
NM_022832.3 | NP_073743.2 | 9 | 2490 | |||
XM_017008557.1 | XP_016864046.1 | 9 | 2515 | |||
GenBank mRNA | AK022614.1 | - | 9 | 2310 | ||
AK024318.1 | - | 9 | 2431 | |||
BC037574.1 | - | 9 | 2439 | 5529667 |