Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024047.4 | NP_076952.1 | 3 | 742 | ||
NM_198038.2 | NP_932155.1 | 3 | 384 | |||
XM_011532031.2 | XP_011530333.1 | 3 | 745 | |||
XM_011532032.2 | XP_011530334.1 | 3 | 2921 | |||
GenBank mRNA | AF265341.1 | - | 3 | 383 | ||
AF273028.1 | - | 3 | 706 | |||
AK027295.1 | - | 3 | 732 | |||
AK074845.1 | - | 3 | 384 | |||
AK075408.1 | - | 3 | 662 | |||
AY026252.1 | - | 3 | 737 | |||
AY359123.1 | - | 3 | 436 | |||
BC000542.2 | - | 3 | 502 | 3163944 | ||
BI552154.1 | - | 382 |