Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM57A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318006.1 | NP_001304935.1 | 3 | 461 | ||
NM_024792.2 | NP_079068.1 | 3 | 461 | |||
GenBank mRNA | AF477201.1 | - | 3 | 449 | ||
AK025935.1 | - | 3 | 400 | |||
AK125146.1 | - | 2 | 666 | |||
AK292065.1 | - | 3 | 400 | |||
BC026023.1 | - | 3 | 423 | 4548729 |