Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROSER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025138.4 | NP_079414.3 | 13 | 3922 | ||
XM_005266545.3 | XP_005266602.1 | 12 | 3895 | |||
XM_011535239.2 | XP_011533541.1 | 12 | 3855 | |||
GenBank mRNA | AB107354.1 | - | 13 | 3922 | ||
AL833854.1 | - | 3 | 1649 | |||
BC064348.1 | - | 13 | 3177 | 4939198 | ||
BX647471.1 | - | 13 | 3344 |