Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278919.1 | NP_001265848.1 | 12 | 2557 | ||
NM_001278920.1 | NP_001265849.1 | 12 | 2424 | |||
NM_030779.3 | NP_110406.1 | 13 | 2665 | |||
NM_173092.2 | NP_775115.1 | 13 | 2398 | |||
XM_011525308.2 | XP_011523610.1 | 13 | 2665 | |||
XM_011525309.2 | XP_011523611.1 | 13 | 2665 | |||
XM_011525310.2 | XP_011523612.1 | 13 | 2398 | |||
XM_017025175.1 | XP_016880664.1 | 13 | 2665 | |||
XM_017025176.1 | XP_016880665.1 | 13 | 2632 | |||
XM_017025177.1 | XP_016880666.1 | 12 | 2557 | |||
XM_017025178.1 | XP_016880667.1 | 12 | 2524 | |||
XM_017025179.1 | XP_016880668.1 | 14 | 2506 | |||
XM_017025180.1 | XP_016880669.1 | 13 | 2501 | |||
GenBank mRNA | AF311913.1 | - | 13 | 2596 | ||
AK090969.1 | - | 6 | 895 | |||
AK091877.1 | - | 13 | 2398 | |||
AK296498.1 | - | 12 | 2530 | |||
AK298363.1 | - | 12 | 2399 |