Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324512.1 | NP_001311441.1 | 5 | 1201 | ||
NM_030955.3 | NP_112217.2 | 5 | 1201 | |||
XM_017009905.1 | XP_016865394.1 | 5 | 1189 | |||
XM_017009906.1 | XP_016865395.1 | 4 | 590 | |||
GenBank mRNA | AJ250725.1 | - | 5 | 1191 | ||
BC131733.1 | - | 5 | 950 | 40124828 |