Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282352.1 | NP_001269281.1 | 7 | 880 | ||
NM_030957.3 | NP_112219.3 | 20 | 2691 | |||
XM_006722917.3 | XP_006722980.1 | 19 | 2537 | |||
XM_017027338.1 | XP_016882827.1 | 19 | 2547 | |||
XM_017027339.1 | XP_016882828.1 | 10 | 1171 | |||
XM_017027340.1 | XP_016882829.1 | 9 | 1031 | |||
GenBank mRNA | AB209515.1 | - | 19 | 2527 | ||
AF163762.1 | - | 17 | 2307 | |||
AK131414.1 | - | 7 | 880 | |||
BC029308.1 | - | 1 | 38 |