Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LONP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300948.1 | NP_001287877.1 | 2 | 637 | ||
NM_031490.3 | NP_113678.2 | 2 | 637 | |||
XM_017023755.1 | XP_016879244.1 | 2 | 504 | |||
XM_017023756.1 | XP_016879245.1 | 2 | 502 | |||
GenBank mRNA | AJ548761.1 | - | 2 | 504 | ||
AK027544.1 | - | 2 | 468 | |||
AK074775.1 | - | 2 | 502 | |||
AK127125.1 | - | 3 | 400 | |||
AK294779.1 | - | 2 | 449 | |||
BC093910.1 | - | 2 | 471 | 7939755 | ||
BC093912.1 | - | 2 | 471 | 7939757 | ||
BC143246.1 | - | 2 | 471 | 9051747 |