Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317395.1 | NP_001304324.1 | 2 | 379 | ||
NM_001317399.1 | NP_001304328.1 | 2 | 379 | |||
NM_031954.4 | NP_114160.1 | 2 | 379 | |||
NR_133897.1 | 2 | 379 | ||||
GenBank mRNA | AF113208.1 | - | 2 | 124 | ||
AK027543.1 | - | 2 | 185 | |||
AK056912.1 | - | 2 | 1784 | |||
AK092170.1 | - | 2 | 168 | |||
AK222548.1 | - | 2 | 135 | |||
AY205299.1 | - | 2 | 185 | |||
AY597809.1 | - | 1 | 97 | |||
BC040062.1 | - | 2 | 132 | 5494016 | ||
BX333975.2 | - | 2 | 129 | |||
DA440672.1 | - | 2 | 379 |