Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042403.2 | NP_001035862.1 | 2 | 434 | ||
NM_001278393.1 | NP_001265322.1 | 2 | 650 | |||
NM_032147.4 | NP_115523.2 | 2 | 579 | |||
NR_125360.1 | 1 | 360 | ||||
XM_005269172.1 | XP_005269229.1 | 2 | 573 | |||
XM_005269173.1 | XP_005269230.1 | 2 | 650 | |||
XM_005269174.1 | XP_005269231.1 | 2 | 417 | |||
XM_011538800.2 | XP_011537102.1 | 3 | 600 | |||
XM_011538801.1 | XP_011537103.1 | 3 | 460 | |||
XM_011538802.1 | XP_011537104.1 | 2 | 420 | |||
XM_011538803.1 | XP_011537105.1 | 2 | 425 | |||
XM_011538804.2 | XP_011537106.1 | 2 | 633 | |||
XM_011538805.1 | XP_011537107.1 | 2 | 1274 | |||
XM_011538806.2 | XP_011537108.1 | 2 | 573 | |||
XM_017020009.1 | XP_016875498.1 | 2 | 568 | |||
XM_017020010.1 | XP_016875499.1 | 2 | 420 | |||
XM_017020011.1 | XP_016875500.1 | 3 | 446 | |||
XM_017020012.1 | XP_016875501.1 | 2 | 573 | |||
GenBank mRNA | AK027434.1 | - | 2 | 385 | ||
AK315697.1 | - | 2 | 582 | |||
AL136825.1 | - | 2 | 540 | |||
BC030704.1 | - | 2 | 415 | 4825887 | ||
BI458616.1 | - | 2 | 586 | |||
DA092275.1 | - | 2 | 391 | |||
DB462518.1 | - | 2 | 434 |