Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACAD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032169.4 | NP_115545.3 | 6 | 1185 | ||
NR_132426.1 | 6 | 1185 | ||||
NR_132427.1 | 6 | 1185 | ||||
NR_132428.1 | 6 | 1185 | ||||
GenBank mRNA | AJ608287.1 | - | 6 | 1136 | ||
AK131265.1 | - | 8 | 982 | |||
AK308380.1 | - | 7 | 775 | |||
AL832873.1 | - | 6 | 1136 | |||
AL833721.1 | - | 6 | 703 | |||
AX803783.1 | - | 6 | 1042 | |||
AX803785.1 | - | 6 | 805 | |||
BC019607.1 | - | 1 | 81 | |||
BC125204.1 | - | 6 | 1120 | 40123454 | ||
BC125205.1 | - | 6 | 1120 |
Gene Symbol: NPHP3-ACAD11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037804.1 | 31 | 4646 | |||
GenBank mRNA | AB082531.1 | - | 31 | 4646 |