Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPPL2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032802.3 | NP_116191.2 | 10, 11 | 1263 | ||
XM_005254722.3 | XP_005254779.1 | 10, 11 | 1262 | |||
XM_017022680.1 | XP_016878169.1 | 10, 11 | 1262 | |||
XM_017022681.1 | XP_016878170.1 | 10, 11 | 1262 | |||
GenBank mRNA | AJ345028.1 | - | 10, 11 | 1241 | ||
AJ420896.1 | - | 8, 9 | 814 | |||
AK027446.1 | - | 8, 9 | 815 | |||
AK075454.1 | - | 10, 11 | 1252 | |||
AK315651.1 | - | 10, 11 | 1230 | |||
AY169314.1 | - | 10, 11 | 1259 | |||
BC025740.1 | - | 10, 11 | 1226 | 5223715 |