Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNH7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033272.3 | NP_150375.2 | 2 | 458 | ||
NM_173162.2 | NP_775185.1 | 2 | 458 | |||
XM_011512109.2 | XP_011510411.1 | 2 | 348 | |||
XM_017005218.1 | XP_016860707.1 | 2 | 348 | |||
XM_017005219.1 | XP_016860708.1 | 2 | 348 | |||
XM_017005220.1 | XP_016860709.1 | 2 | 348 | |||
XM_017005221.1 | XP_016860710.1 | 2 | 348 | |||
GenBank mRNA | AF032897.1 | - | 2 | 329 | ||
BC035815.1 | - | 2 | 441 | 5769938 | ||
DR002425.1 | - | 2 | 463 |