Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTRA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053044.4 | NP_444272.1 | 9 | 2076 | ||
GenBank mRNA | AY040094.2 | - | 9 | 2086 | ||
AY280665.1 | - | 9 | 2064 | |||
BC034390.1 | - | 9 | 2076 | 5180585 | ||
BC035717.1 | - | 9 | 2076 | 5588608 |