Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127441.1 | NP_001120913.1 | 8 | 1093 | ||
NM_001127442.1 | NP_001120914.1 | 6 | 982 | |||
NM_001318223.1 | NP_001305152.1 | 6 | 982 | |||
NM_080385.4 | NP_525124.3 | 7 | 1040 | |||
XM_005250710.1 | XP_005250767.1 | 7 | 1021 | |||
XM_005250712.1 | XP_005250769.1 | 6 | 935 | |||
XM_011516698.1 | XP_011515000.1 | 6 | 1658 | |||
XM_011516701.1 | XP_011515003.1 | 6 | 1575 | |||
XM_011516702.1 | XP_011515004.1 | 6 | 1561 | |||
XM_011516703.1 | XP_011515005.1 | 6 | 1554 | |||
XM_011516704.1 | XP_011515006.1 | 6 | 1672 | |||
XM_011516705.1 | XP_011515007.1 | 6 | 1672 | |||
GenBank mRNA | AF384667.1 | - | 5 | 473 | ||
AK093288.1 | - | 6 | 695 | |||
AK131191.1 | - | 6 | 918 | |||
AK315256.1 | - | 6 | 956 | |||
AY155229.1 | - | 7 | 993 | |||
AY155230.1 | - | 8 | 1063 | |||
BC039362.1 | - | 8 | 1067 | 5270617 | ||
BC042996.1 | - | 6 | 1609 | 5296768 |