Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MITD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320417.1 | NP_001307346.1 | 5, 6 | 794 | ||
NM_001320418.1 | NP_001307347.1 | 5, 6 | 670 | |||
NM_001320419.1 | NP_001307348.1 | 4, 5 | 583 | |||
NM_138798.2 | NP_620153.1 | 5, 6 | 670 | |||
XM_011510581.2 | XP_011508883.1 | 5, 6 | 682 | |||
XM_011510582.2 | XP_011508884.1 | 5, 6 | 682 | |||
XM_011510583.1 | XP_011508885.1 | 5, 6 | 768 | |||
XM_017003314.1 | XP_016858803.1 | 4, 5 | 595 | |||
XM_017003315.1 | XP_016858804.1 | 5, 6 | 682 | |||
GenBank mRNA | AL161992.1 | - | 3, 4 | 2919 | ||
BC018453.1 | - | 5, 6 | 638 | 4509694 | ||
BG504564.1 | - | 4, 5 | 574 | |||
BU507044.1 | - | 5, 6 | 652 |