Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH5A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080.3 | NP_001071.1 | 10 | 2170 | ||
NM_170740.1 | NP_733936.1 | 11 | 2209 | |||
GenBank mRNA | AJ427354.2 | - | 11 | 1977 | ||
AJ427355.2 | - | 10 | 2142 | |||
Y11192.1 | - | 10 | 2141 |