Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002238.3 | NP_002229.1 | 3 | 397 | ||
NM_172362.2 | NP_758872.1 | 3 | 397 | |||
GenBank mRNA | AF078741.1 | - | 3 | 263 | ||
AF078742.1 | - | 3 | 263 | |||
AJ001366.1 | - | 3 | 397 | |||
BC113709.1 | - | 3 | 297 | 8322761 | ||
BC143599.1 | - | 3 | 298 | 9052111 | ||
KT895501.1 | - | 3 | 227 | |||
KT895502.1 | - | 3 | 227 |