Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002412.4 | NP_002403.2 | 2 | 211 | ||
XM_005252682.2 | XP_005252739.1 | 2 | 248 | |||
GenBank mRNA | AK293593.1 | - | 2 | 181 | ||
BC000824.1 | - | 2 | 139 | 3454207 | ||
BI520029.1 | - | 2 | 193 | |||
BT006714.1 | - | 1 | 86 | |||
DC387686.1 | - | 2 | 205 | |||
DC416206.1 | - | 2 | 211 | |||
KC249995.1 | - | 1 | 120 | |||
M29971.1 | - | 2 | 182 | |||
M31767.1 | - | 1 | 108 | |||
M60761.1 | - | 1 | 101 | |||
X54228.1 | - | 1 | 126 |