Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCSK9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174936.3 | NP_777596.2 | 7 | 1500 | ||
NR_110451.1 | 5 | 797 | ||||
GenBank mRNA | AK075365.1 | - | 7 | 1429 | ||
AK122717.1 | - | 8 | 1185 | |||
AK124635.1 | - | 4 | 1862 | |||
AK297473.1 | - | 5 | 797 | |||
EF692496.1 | - | 7 | 1138 |