Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPPL2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175882.2 | NP_787078.2 | 1 | 565 | ||
GenBank mRNA | BC022041.1 | - | 1 | 567 | 4825062 | |
BC025401.1 | - | 1 | 570 | 4829763 |