Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZADH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306093.1 | NP_001293022.1 | 2 | 802 | ||
NM_175907.5 | NP_787103.1 | 2 | 980 | |||
GenBank mRNA | AK292880.1 | - | 2 | 735 | ||
AK302801.1 | - | 2 | 532 | |||
BC018081.1 | - | 2 | 958 | |||
BC033780.1 | - | 2 | 866 | 4510603 | ||
BC078661.1 | - | 2 | 793 | 5575156 | ||
BX647730.1 | - | 2 | 804 |