Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144072.1 | NP_001137544.1 | 9 | 2557 | ||
NM_177967.3 | NP_808882.1 | 7 | 2471 | |||
NR_026644.1 | 9 | 2701 | ||||
XM_006719948.3 | XP_006720011.1 | 10 | 2267 | |||
XM_011521082.2 | XP_011519384.1 | 7 | 2327 | |||
XM_011521083.2 | XP_011519385.1 | 8 | 2034 | |||
XM_011521084.2 | XP_011519386.1 | 9 | 2122 | |||
XM_017020553.1 | XP_016876042.1 | 4 | 5349 | |||
GenBank mRNA | AF339812.1 | - | 4 | 1541 | ||
AK054563.1 | - | 7 | 2472 | |||
AK055110.1 | - | 6 | 2254 | |||
AK075419.1 | - | 9 | 2128 | |||
AK075516.1 | - | 7 | 1898 | |||
AK124404.1 | - | 9 | 1855 | |||
AK129861.1 | - | 10 | 1552 | |||
BC063559.1 | - | 7 | 1916 | |||
BC072674.1 | - | 5 | 1693 |