Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AASDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286668.1 | NP_001273597.1 | 11 | 2556 | ||
NM_001286669.1 | NP_001273598.1 | 11 | 2707 | |||
NM_001286670.1 | NP_001273599.1 | 9 | 2231 | |||
NM_001323890.1 | NP_001310819.1 | 13 | 2945 | |||
NM_001323899.1 | NP_001310828.1 | 11 | 2721 | |||
NM_181806.3 | NP_861522.2 | 12 | 2828 | |||
XM_017007740.1 | XP_016863229.1 | 12 | 2789 | |||
XM_017007741.1 | XP_016863230.1 | 13 | 2904 | |||
XM_017007742.1 | XP_016863231.1 | 10 | 2604 | |||
XM_017007744.1 | XP_016863233.1 | 9 | 2411 | |||
GenBank mRNA | AF516672.1 | - | 11 | 2471 | ||
AK294332.1 | - | 9 | 2193 | |||
AK304309.1 | - | 12 | 2819 | |||
AK316296.1 | - | 12 | 2809 | |||
AY314787.1 | - | 12 | 2791 | |||
AY422212.1 | - | 12 | 2743 | |||
BX640635.1 | - | 7 | 3659 |