Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHRS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005794.3 | NP_005785.1 | 9 | 1594 | ||
NM_182908.4 | NP_878912.1 | 9 | 1605 | |||
XM_005267249.1 | XP_005267306.1 | 9 | 1306 | |||
XM_006720001.3 | XP_006720064.1 | 9 | 1269 | |||
XM_011536338.1 | XP_011534640.1 | 8 | 1235 | |||
XM_011536339.2 | XP_011534641.1 | 8 | 1085 | |||
GenBank mRNA | AB209653.1 | - | 7 | 5236 | ||
AK000345.1 | - | 1 | 2513 | |||
AK092834.1 | - | 5 | 3005 | |||
AK222857.1 | - | 9 | 1594 | |||
AK309945.1 | - | 9 | 3081 | |||
BC002786.2 | - | 9 | 1596 | 3614599 | ||
BC007339.2 | - | 8 | 1120 |