Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318781.1 | NP_001305710.1 | 12 | 1936 | ||
NM_182920.1 | NP_891550.1 | 13 | 2020 | |||
GenBank mRNA | AB037733.1 | - | 12 | 1515 | ||
AF261918.1 | - | 13 | 2000 | |||
AF488803.1 | - | 13 | 2020 | |||
BC130578.1 | - | 13 | 2010 | 40146609 | ||
BC171764.1 | - | 12 | 1926 | 9054417 |